Bulk Dispensers with Lid or Open Top, S-Curve
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Multiple uses for gloves, face masks, hair nets, bouffants, shoe covers, and more.
- Available in 1/4" clear acrylic or 1/4" clear PETG material
- Flame treated edges for a smooth finish
- Includes wall mounting bracket
Rubber pads on bottom corners to prevent slipping and scratching.
Spécifications
- Catalog No:
- 10828-392
- 10693-128
- 10693-126
- 10693-130
- 10693-134
- 10693-132
- 10828-390
- 10693-136
- PPE type:
- Multi-use
- Multi-use
- Multi-use
- Multi-use
- Multi-use
- Multi-use
- Multi-use
- Multi-use
- Material:
- PETG
- PETG
- Acrylic
- PETG
- Acrylic
- PETG
- Acrylic
- Acrylic
- No. of compartments:
- 3
- 1
- 1
- 2
- 2
- 3
- 2
- 3
- Dispenser type:
- Wall-mountable
- Wall-mountable
- Wall-mountable
- Wall-mountable
- Wall-mountable
- Wall-mountable
- Wall-mountable
- Wall-mountable