GTS Rx Shipper Family, Cold Chain Technologies
Fournisseur: Cold Chain Technologies
Certificats
A propos de cet article
Pre-qualified for multiple temperature ranges: 2 to 8 °C for 36+ hours; room temperature (15 to 25°C or 10 to 30°C) for 1 to 2 days; frozen for 36+ hours.
- Single refrigerant SKU across all sizes reduces inventory
- Training videos & education material available
- Compliance documentation for regulatory & accreditation audits
- Performance Qualification (PQ) study kit and service available
- Nationwide delivery available via truckload, LTL, or parcel service
- Specifically qualified for specialty pharmacies and last mile shipments against ISTA 7E ambient profiles
Shipment of high value, temperature sensitive therapies requires uniquely designed packaging and validated processes from start to finish. Recommended uses include overnight domestic shipments, specialty and mail-order pharmacies, wholesale distributors, and drug sampling programs.
Customers Who Bought This Also Bought