Removable Cryogenic Labels for Laser Printers, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
TransCryo-LazrTAG™ laser printable cryogenic labels are designed for the long-term storage in liquid nitrogen and deep-freezing harsh environments for biomedical research and clinical laboratories.
- Transparent, water-resistant
- Wear and tear proof
- Do not jam printers
- Resistant to UV and gamma irradiation
- Easy to format
- Resistant to autoclave and boiling water
- Withstand a wide temperature range (–196 to 120 °C)
Cryo labels accept desktop laser printing and permanent marker. The sheet size measures about 216×279.4 mm (8.5×11").
Customers Who Bought This Also Bought