Cryo-ID-Kit™ Tags, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
These tags are used for identification of metal racks and other containers for long-term storage in liquid nitrogen and ultra-low temperature freezers.
- Suitable temperature ranges from –196 to +80 °C
Labels can be printed with all major brands of thermal transfer barcode printers (SATO, Zebra, Intermec, Printronix, Datamax-O’Neil, CAB, and more) or inscribed with permanent markers such as our cryo-markers.
Delivery: Includes 12 plastic Cryo-Cards™ with connection hooks and thermal transfer cryo labels.
Customers Who Bought This Also Bought