Photo-etched Cover Slips, Standard and German Glass, Bellco Glass
Fournisseur: Bellco Glass
Certificats
A propos de cet article
Schott D236 glass ideal for fluorescence and confocal microscopy.
- Diamond grid pattern
12 mm round coverslip is manufactured from #1 thickness (0.13 to 0.16 mm) Schott D263 german glass and is ideal for fluorescence and confocal microscopy. The coverslip has 100 and 520 alphanumeric locations in a diamond pattern and each measures 0.6×0.6 mm with line thickness of 0.02 mm. This coverslip fits into 24 Well microplates.
Customers Who Bought This Also Bought