Serological Pipettes, Bellco Glass
Fournisseur: Bellco Glass
Certificats
A propos de cet article
Type 1 Class B Borosilicate Glass.
- Calibrated Tip
Plain top promotes rapid even flow with a calibrated tip. All Bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.
Customers Who Bought This Also Bought