Horizon Eneregy Monitor
# de catalogue 470314-474
Fournisseur: HORIZON FUEL CELL TECHNOLOGIES
Spécifications
- Description:Data Logging Energy Monitor
- Cat. no.:470314-474
Spécifications
A propos de cet article
Compatible with any of Horizon's science education kits.
- Windows and mac compatible
- Use with or without a computer
- Export data to excel or CSV format
- Record videos for playback and review
Voltage, current, power, joules, resistance and even RPM speed available at the touch of a button—displayed either on the LCD screen of the monitor itself or as graphical data on your PC. The Horizon Energy Monitor helps bring lab and field experiments back into the classroom, making it the ideal tool to complement the wind to hydrogen science Kit (FCJJ-56), the renewable energy science kit (FCJJ-37) and the wind energy science kit (FCJJ-39).
Certifications: CoC, RoHS, EN 71:PART1;PART2;PART3, EN 62115, PHTH-EU, ASTM F963, CPSIA-LEAD, CPSIA-PHTHALATES, REACH, CA
Customers Who Bought This Also Bought