Vikan® Long Handle Utility Brush, Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This long handled wash brushes reaches into parts of equipment that are hard to access. The angled bristles are stiff, creating a washing action.
- Stiff polyester bristles
- Polypropylene block
- Dishwasher safe
- Autoclavable up to 250°F
Certifications: Meet FDA standards.
Customers Who Bought This Also Bought