ContactUsLinkComponent
Wound Closure Strips, DUKAL Corporation®
Wound Closure Strips, DUKAL Corporation®
# de catalogue 76321-916
Fournisseur:  Dukal
Strips Wound Closure St 1/4X3In BX150
Wound Closure Strips, DUKAL Corporation®
# de catalogue 76321-916
Fournisseur:  Dukal
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Dimensions:
    0.25×3"
  • Sterility:
    Sterile
  • Color:
    White
  • Description:
    Wound Closure Strips
  • Cat. no.:
    76321-916
Spécifications
A propos de cet article
Keeps wounds closed without significant tension, edema, stretch, or movement.
  • Provides support following early suture/staple removal
  • Translucent and porous
  • Not made with natural rubber latex
Adhesive backed, thin non-woven, reinforced fibers are arranged in a bilateral pattern for high tensile strength.
Packaging: Strips are packaged 3 per pack, 50 packs per box, 4 boxes per case.
Customers Who Bought This Also Bought