Material Rolls for Material Dispenser with Liner Remover, Brady
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
Thermal transfer white labels for the Brady® Material dispenser with liner remover.
- Sticks to any clean, dry surface
- Ideal for use in wire and cable identification, or temporary vial labeling
- Recommended for indoor use
The B-402 material completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind. Ideal for use in laboratory settings, asset tracking, or general use. Easy removal saves time during re-labeling processes. Pressure-sensitive adhesive allows for quick and easy application.
Permanent or repositionable once applied, B-498 CleanLift® series vinyl cloth is there to stay, unless you want to change it. The adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning.
Nylon cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces.
Customers Who Bought This Also Bought