Vikan Nylon Floor Scraper, Remco
# de catalogue 76006-598
Fournisseur: Remco Products
Spécifications
- Dimensions:10.25L×1.25W×6.90"H
- Cat. no.:76006-598
- Weight:0.65 lbs.
Spécifications
A propos de cet article
Floor scraper with heavy-duty nylon blade that do not damage floors or machines.
- High-quality floor scraper
- Nylon floor scraper
Certifications: Meet FDA standards.
Customers Who Bought This Also Bought