3200 Series Premium 2" Fabric Upholstered Double Hinge Folding Chairs, National Public Seating
Fournisseur: National Public Seating
Certificats
A propos de cet article
The 3200 series has brought the potential comfort of folding chair to a whole new level with a super-soft 2" thick cushion on a masonite board with 8 rivets.
- Fabric and foam are cal 117 rated, Scotchguarded fabric is rated to 30000 double rubs
- Padded backs are fully covered in fabric on both sides
- Two U-shaped double riveted cross braces - one in front and one in back
- Institutional-grade powder-coated frame is made from heavy-duty powder-coated 18-gauge round tubing
- Full-size, double-contoured backrest, upholstered and firmly secured to the frame by six rivets and waterfall seat
- Double hinges on each side for added stability and durability
- Color coordinated polypropylene glides and v-tip plugs for added stability
Institutional grade fabric over 1.25", 0.9 lbs. density foam seat and 1.2 lbs. density back
10 year warranty
Certifications: MAS Certified Green, ANSI/BIFMA X7.1-2007
Spécifications
- Catalog No:
- 76276-414
- 76276-412
- 76276-410
- 76276-408
- 76276-406
- 76276-404
- Model:
- 76276-414
- 76276-412
- 76276-410
- 76276-408
- 76276-406
- 76276-404
- Frame material:
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Seat/back color:
- Gold
- Burgundy
- Blue
- Charcoal
- Black
- Beige
- Seat/back material:
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Seat dimensions:
- W15.75×D15.5×H18.5"
- W15.75×D15.5×H18.5"
- W15.75×D15.5×H18.5"
- W15.75×D15.5×H18.5"
- W15.75×D15.5×H18.5"
- W15.75×D15.5×H18.5"
- Backrest dimensions:
- W18.75×H8.75"
- W18.75×H8.75"
- W18.75×H8.75"
- W18.75×H8.75"
- W18.75×H8.75"
- W18.75×H8.75"
- Weight capacity:
- 480 lbs.
- 480 lbs.
- 480 lbs.
- 480 lbs.
- 480 lbs.
- 480 lbs.