CE Multisurface Cleaner™, Cleaning Mop, Vileda Professional®
# de catalogue 76318-342
Fournisseur: Vileda Professional - FHP
Spécifications
- Description:CE Multisurface Cleaner™
- Material:Plastic/Metal
- Size:23"
- Cat. no.:76318-342
Spécifications
A propos de cet article
Designed to clean pipes, hoods, and other hard to reach areas or uneven surfaces with minimal effort and maximum efficiency. Versatile and easily bendable to give the tool different shapes to effectively clean any surface.
- Completely autoclavable and rust free
- Extremely versatile
- Easily bendable to a variety of different shapes to effectively clean any surface
- Sturdy enough to not bend in use
Customized tools for special surfaces. The easiest, most effective way to clean pipes, soft walls curtains, tanks and other hard to reach areas in Controlled Environments.
A new line of specialty above-the-floor tools by Vileda Professional®. This line is ideal for all surfaces that are hard or difficult to reach, clean, and disinfect. Specialty tools use Controlled Environment compatible disposable microfiber sleeves that are already validated materials and are an extension of a current CE floor cleaning system.
Certifications: Meet CE requirements up to ISO 4-9/GMP A/B, C, D.
Customers Who Bought This Also Bought