Graduated Glass Gas Tubes
Fournisseur: Wards
Certificats
Certificats
A propos de cet article
Made of borosilicate glass.
Customers Who Bought This Also Bought
Potassium dihydrogen phosphate ≥99.0% ACS, VWR Chemicals BDH®
Fournisseur: VWR International
360,25 $ to 1 246,54 $
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
60,20 $ to 3 424,99 $
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
214,30 $ to 2 349,04 $
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
155,16 $
Methanol ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Fournisseur: MilliporeSigma
449,31 $ to 1 761,78 $
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
72,65 $ to 846,50 $