Sapphire Deep Well Plates, Azer Scientific
Fournisseur: Azer Scientific Inc.
Certificats
A propos de cet article
Sapphire deep well plates meet SBS dimensional standards for use in all common thermocyclers.
- Raised wells, U-shaped bottom
- Resistant to extreme temperatures
- Equipped with raised alphanumeric grid
Highly chemical resistant deep well plates are suited for the most sensitive diagnostic applications. Raised wells reduce cross contamination and allow for secure sealing with heat or adhesive sealing mats. Both the 1mL round-well plate and 2mL square-well plate can withstand freezing and microwave desfrosting.
Packaging: Plates are packaged 20 per pack.
Customers Who Bought This Also Bought