Borosilicate Glass Burettes with Pinch Clips
Fournisseur: Wards
Certificats
A propos de cet article
Single bore burettes with detachable rubber tube and pinch clip.
- Made of high quality 3.3 borosilicate glass
- 0.10 ml sub divisions
- Tolerance of ±0.20 ml
Certifications: DIN ISO 385 standard, Class B
Customers Who Bought This Also Bought