Glass Microscope Slides, Azer Scientific
Fournisseur: Azer Scientific Inc.
Certificats
A propos de cet article
General-purpose microscope slides offer quality and value for any laboratory.
- Extra white soda-lime glass with 90 ° corners
- Manufactured in Germany
Choose any one of these general-purpose microscope slides for clinical, environmental, industrial, or other routine microscopy work. Extra white soda-lime glass will not interfere with fluorescent staining procedures. Uniform 1.0 mm thickness makes these sturdy enough for field work.
Packaging: Slides are packaged 72 per box, 20 boxes per case.
Customers Who Bought This Also Bought