Dispenser for Sterile/Pair-Packaged Gloves, 3 Compartments, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Glove dispensers with 3 compartment designed for sterile or pair-packaged gloves.
- Front access
- Flame polished edges for smoothness
Wall mounts or free-standing.
Each compartment measures 6.5W×12" D
Customers Who Bought This Also Bought