Ball Mills, PTA, Nidec Shimpo
Fournisseur: Nidec Shimpo America
Certificats
A propos de cet article
Compact design that is ideal for small-scale production and is constructed using powder-coated steel for long-lasting durability.
- Easy adjustments can be made to the sturdy roller bars to accommodate jars from 1 to 10 l
- Comes standard with a breaker switch
Heavy-duty ball milling machines enable very precise milling, in porcelain jars, even for relatively tough materials, wet or dry.
Spécifications
- Catalog No:
- 76307-538
- 76307-536
- Motor power:
- 180 W
- 180 W
- Torque:
- ~5 Nm
- ~5 Nm