Sterile Microscope Slides, Azer Scientific
# de catalogue 76303-034
Fournisseur: Azer Scientific Inc.
Spécifications
- Description:Microscope slides, sterile
- L×W:75×25 mm
- Thickness:1.0 mm
Spécifications
A propos de cet article
Sterilized microscope slides feature a single frosted end marking area.
- Individually wrapped
Single frosted end, sterile microscope slides are suitable for use in clinical settings when direct specimen mounting is required for immediate staining and examination.
Packaging: Slides are packaged 50 per pack.
Customers Who Bought This Also Bought