Ergolux Jr. Lab Stools
Fournisseur: BevCo Precision Manufacturing
Certificats
A propos de cet article
Advanced stress relief With the Ergolux Jr. series.
- The 2.5" thick ultrasoft, high-density, visco-elastic memory foam seat provides resilient, long-lasting ergonomic support
- The 14.5" diameter seat is covered by puncture, water, and stain-resistant polyurethane
- Stool is non-slip and easy-to-clean
- Easy-to-use pneumatic height adjustment
- Sturdy 25" diameter black nylon base
- Dual-wheel hard floor casters
- Black seat color
- 300 lb. weight capacity
- For standard environments
The stress-relieving qualities of the stool are heightened because they respond to your body's unique shape, weight, and temperature. The Ergolux Jr. series is ideal for a wide range of applications, including general office, medical, laboratory, and higher education applications.
Certifications: ANSI/BIFMA; GreenGuard Gold.
Ordering information: 12-year warranty.
Spécifications
- Catalog No:
- 76319-094
- 76319-098
- 76319-120
- 76319-124
- Model:
- Environmentally Preferable:
- Safer Human and Environmental Health
- Frame colour/finish:
- Black
- Frame material:
- 25" nylon
- Seat/back color:
- Black
- Seat dimensions:
- Ø14.5×T2.5"
- Environmental certification:
- Greenguard GOLD