Nickel Crucibles with Lids
Fournisseur: Wards
Certificats
Certificats
A propos de cet article
Easy to clean nickel crucibles work with temperatures up to 1000 °C.
Customers Who Bought This Also Bought
Nalgene® Cryoware Marker Set, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
$142.56 to $406.56
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
$881.47
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Fournisseur: VWR International
$314.91
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$209.41 to $1,276.12
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$464.49
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
$534.61 to $1,523.31