LOCTITE® GRD C (084) Thread lockers, Henkel Corporation
# de catalogue 76313-916
Fournisseur: Loctite
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Volume:50 ml
- Description:Thread Lockers
- Cat. no.:76313-916
Spécifications
A propos de cet article
LOCTITE® Letter Grade C (084) Thread lockers are used to lock and seal fine threaded nuts, bolts, and studs in a wide variety of applications.
- Medium strength
These are removable with tools.
Ordering information: Ten pieces per case. Must be ordered in case quantity.
Note: This is a regional approval. Please contact a local Technical Service Center for more information and clarification.
Note: This is a regional approval. Please contact a local Technical Service Center for more information and clarification.
Packaging: 50 ml bottle, 0.077 kg Package Weight, 5.9L×2.9W×15.9H cm Packaging Dimension.
Customers Who Bought This Also Bought