Utility Brush, Toothbrush-Style, Justman
# de catalogue 30620-638
Fournisseur: Justman Brush
Spécifications
- Bristle Color:White
- Bristle Material:Nylon
- Handle Material:Plastic
- Total length:7 1/4"
- Cat. no.:30620-638
- No. of Rows:3×7
- Handle Color:Black
- Supplier No.:710478
- Brush Head Length:1 1/2"
- Trim Length:½"
Spécifications
A propos de cet article
Utility brush has a toothbrush style, with stiff white nylon bristles in an array of 3×7 rows.
Customers Who Bought This Also Bought