Mohr's Style Burettes
Fournisseur: GSC International
Certificats
A propos de cet article
These borosilicate glass burettes have a reinforced opening and are permanently graduated with subdivisions.
Tip set, 470313-342 attaches to Mohr's-style burettes. It includes 2" of latex tubing, a pinch clamp, and a tip made from capillary glass.
Customers Who Bought This Also Bought