A propos de cet article
High-quality science lab table, high pressure laminate top, solid wood legs table meet MAS certified green standards and are a great value.
- Solid ash legs and apron
- NEMA standard compliant black high-pressure laminate top on a 11/8" thick core
- Legs attach with metal on metal double bolt through assembly on 13-gauge steel corner braces
- Adjustable glides for accurate floor leveling
- Rubber liquid-protecting leg boots
- Top surface is resistant to scratches, boiling water, high temperatures and acid stains
- Ten year warranty
National public seating strong and sturdy science tables are a necessity for the science lab. Built from solid ash wood with an attractive polyurethane clear finish, our high pressure laminate science lab tables are perfect for many science and chemistry classrooms. HPL tabletops provide an exceptional work surface and are made with a high-pressure laminate surface that has been tested according to NEMA standard testing to resist scratches, boiling water, high temperatures, and many forms of stains including a number of acids. Also exceeds 1800 wear cycles and over 125" ball impact resistance.
HPL tabletops come equipped with our top of the line ribbed-tongue T-molding. The table legs are assembled with a 13 gauge, heavy-duty steel brace attached with a metal-on-metal double bolt connection for superior durability and to provide a no-wobble experience. NPS science tables provides a choice of book compartments or solid front aprons, with an industry top-of-the-line ash wood frame and legs outfitted with rubber liquid-protecting leg boots and adjustable glides for accurate floor leveling.