ContactUsLinkComponent
3B Scientific® Induction Apparatus
Catalogue # 470233-822
Fournisseur:  American 3B Scientific
undefined
3B Scientific® Induction Apparatus
Catalogue # 470233-822
Fournisseur:  American 3B Scientific
Supplier Number:  1022439
Spécifications
  • Description:
    Induction Apparatus
  • Cat. no.:
    470233-822
Spécifications
À propos de cet article
Apparatus for demonstrating the induced voltage in a frame coil.
  • For tabletop, overhead or inclined demonstration
  • Foldable stand
Apparatus for demonstrating the induced voltage in a frame coil that is moved through the magnetic field produced by a magnet plate of limited area or by the rotation of a current-carrying conductor in the magnetic field of the magnet plate. By varying the speed of motion of the frame coil, the direction of motion and the number of turns in the coil, the induction law can be derived experimentally and quantitatively. The transparent design of the magnetic plate and coils means that they can be demonstrated on the overhead projector. An unfoldable support permits inclined setup.
Ordering information: This item does not include a power supply.