3B Scientific® CT Scanner
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
Perform experiments in computed tomography.
- Individual components allow you to purchase just what you need
- CT sample designed for non-uniform Absorption
CT scanner for turning and moving samples for the purpose of generating tomographic images. All turning and movement is carried out by means of stepper motors. While the measurement is taking place, the scanner moves the sample backwards and forwards between the ultrasonic sensors connected to the outside of the sample container in accordance with the CT algorithm. Includes a sample stage for making recordings of suitable objects for observation. The sample stage is immersed in a sample trough. The whole slider is height adjustable so that it is possible for the area of the sample under investigation to be modified.
Customers Who Bought This Also Bought