Bottles, Wide Mouth, HDPE, C&G Containers
Fournisseur: C&G Containers
Certificats
A propos de cet article
HDPE containers are most commonly used for inorganics.
Customers Who Bought This Also Bought
HDPE containers are most commonly used for inorganics.