ContactUsLinkComponent
Component Balance, 3B Scientific
Component Balance, 3B Scientific
# de catalogue 470233-678
Fournisseur:  American 3B Scientific
Component Balance, 3B Scientific
# de catalogue 470233-678
Fournisseur:  American 3B Scientific
Spécifications
  • Description:
    Component Balance
  • Cat. no.:
    470233-678
Spécifications
A propos de cet article
Scales for measuring components with holding mechanism for measuring air resistance and buoyancy of bodies.
Scales for measuring components with holding mechanism for measuring air resistance and buoyancy of bodies from set U8404260. On rod.
Delivery: Includes rod
Customers Who Bought This Also Bought