KleenGuard® G10 Kraken Grip™ Black Nitrile Gloves, Kimberly-Clark
Fournisseur: Kimberly-Clark
Certificats
A propos de cet article
Black nitrile gloves, fully textured on palm and fingers.
- Tire tread-like texture on the outside for grip
- Powder-free moisture control on the inside for comfort
G10 Kraken Grip™ nitrile gloves are ideally suited for applications in fabrication, machining, parts handling, assembly, indoor/outdoor maintenance, and cleaning.
Certifications: Approved for food handling per US FDA. California Prop 65 compliant.
Packaging: Gloves are packaged in a recyclable container box.
Spécifications
- Catalog No:
- 76319-618
- 76319-620
- 76319-682
- 76319-616
- 76319-684
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes