A propos de cet article
High-quality series deluxe vinyl upholstered stack chairs features a concealed double back with four covered metal tabs for a more refined look.
- Steel standard: ASTM A513
- Foam standard: 1.8 lbs. density, 46 ILD
- Made from rugged 7/8" square 18 gauge steel tubing
- Superior frame strength with 5/8” underseat brace and “H” brace
- Waterfall seat, padded with 2" high-density grade a foam with double-stitched box seat
- 10 year warranty
The 9300 Silhouette stack chairs are durable and classy enough to use in the busiest social setting, be it a dining room or corporate banquet hall. It features a strong square 18 gauge steel frame plus an under-seat H-shaped brace for extra support. The seat features supremely comfortable and spacious 2" thick high-density foam padding with institutional grade vinyl upholstery or fabric, and a waterfall-style edge. Plywood core within both the seat and crown-style backrest which has a built-in handle for easy moving and stacking. Fortifying stacking bars and 12 plastic bumpers protect the powder-coated paint finish from scratches when the chair is stacked (up to ten units high.) Available in choice of institutional grade thick 19 ml vinyl solid fabrics rated to 130000 double rubs. Patterned fabrics rated to 67000 double rubs MDF core seat and back, rocker glides, stack 8 to 10 high with DY81 or DY9000 dollies, 300 lbs. weight capacity.
Spécifications
- Catalog No:
- 76276-474
- 76276-472
- 76276-470
- 76276-482
- 76276-480
- 76276-478
- 76276-476
- 76276-468
- 76276-466
- Model:
- 76276-474
- 76276-472
- 76276-470
- 76276-482
- 76276-480
- 76276-478
- 76276-476
- 76276-468
- 76276-466
- Frame colour/finish:
- Black Sandtex
- Black Sandtex
- Silvervein
- Silvervein
- Black Sandtex
- Black Sandtex
- Black Sandtex
- Black Sandtex
- Silvervein
- Seat/back color:
- Natural Chocolatier
- Ebony Black
- Midnight Blue
- Diamond Burgundy
- Diamond Navy
- Natural Blue
- Natural Greystone
- Panther Black
- Midnight Blue