Evaporating Dishes, Flat Form, PTFE, Chemglass
Supplier: Chemglass
Certificates
About this item
Isostatically molded in pure, FDA approved PTFE.
- Totally chemically resistant and inert
- Super smooth internal finish
Flat Form Evaporating Dishes are designed for the evaporation of solids and supernatant fluids.
Customers Who Bought This Also Bought