PTFE Tubing, Microsolv
Fournisseur: MicroSolv Technology Corp
Certificats
A propos de cet article
PTFE tubings are widely used in all low-pressure laboratory applications.
- Micro-pores allow a quick air exchange
- Suitable for degassed and oxygen-sensitive solvents
Durometer (Hardness): D-50
Customers Who Bought This Also Bought