Full Brim Future Hard Hat, PortWest®
# de catalogue 76105-132
Fournisseur: Portwest
Spécifications
- Color:White
- Description:Full Brim Future Hard Hat
- Cat. no.:76105-132
Spécifications
A propos de cet article
Full Brim hard hat.
- Four points nylon harness with wheel adjustable ratchet
- Soft foam sweat band included
- Adjustable 52 to 63 cm (20.4 to 24.8")
Wide profile provides extra protection against sun, glare, rain, debris and non-toxic splashes.
Certifications: EN 397 (−10/+50 °C); ANSI/ISEA Z89.1 TYPE I (Class E).
Customers Who Bought This Also Bought