WHEATON® Battery Powered Crimpers and Decappers, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These battery powered crimpers and decappers fit standard laboratory vials and seals. They come with a long-lasting battery with lithium ion technology, which allows for up to 800 crimps per charge.
- Powerful motor with a fast cycle
- Cushioned ergonomic handle reduces hand fatigue
- Polished crimping jaws provide consistent sealing
- Can be operated while charging
A warning light indicates when recharging is necessary. The tool can be recharged in one to two hours with the universal power supply included.
The electronically adjustable cycle is accurate and reproducible and can be adjusted with the control pad on top of the unit. The tools are ergonomically designed to reduce strain and arm injury associated with many manual crimping tools.
Customers Who Bought This Also Bought