A propos de cet article
These permanent labels are designed for the identification of tubes, vials, boxes, and other plastic or cardboard containers stored in ultra-low temperature laboratory freezers.
- Temperature range: –80 to +110 °C (–112 to +230 °F)
- Ideal for long-term storage in deep-freeze conditions (–80 °C)
- No cure time; can be stored in freezers immediately after application
- No additional lamination necessary
- Labels and printout resist spraying with alcohols and other chemicals
- Printable with all major thermal transfer printer brands
- Available in a large range of sizes, colors, and configurations
These barcode labels are perfect for identifying small Ø vials, microtubes, Falcon tubes, cell culture plates, microplates, and polypropylene and cardboard freezer boxes. These freezer-grade labels are water-proof and tear-proof and will adhere to containers in ultra-low temperature (–80 °C/–112 °F) lab freezers and dry ice without the need for extra lamination.
Available in multiple configurations and sizes, with a permanent adhesive. They are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, as well as wiping with surface cleaning wipes.