NPS® 6300 Series Heavy - Duty Square - Seat Steel Stools, National Public Seating
Fournisseur: National Public Seating
Certificats
A propos de cet article
High-quality Heavy-Duty Square-Seat Steel Stools are a great value.
- Square, contoured hardboard seats with 16 rivets
- 7/8" O-Ø 18-gauge steel tubing
- 5/8" O-Ø foot rings are welded to each leg by four contact points for added rigidity
- Standard seat dimensions: 14.5L×15" W
- Seats with backrest adjust up/down and forward/back
- Height adjustable seats adjust in 1" increments up to an additional 8"
- Ten year warranty
The durable construction and classic design of the 6300 series heavy-duty square-seat stools with backrests are ideal for hands-on educational settings like art rooms, science labs, home economics, or shop classrooms. These reliable stools feature a strong frame of 18-gauge steel tubing securely welded to the seat pan, which is riveted in sixteen places to wide Masonite seats that will not warp, chip, or crack. They provide years of dependable use. Footrests are securely bonded to each leg with four spotwell contact points. The adjustable backrests can be moved up/down and forward/back.
Certifications: MAS Certified Green. ANSI BIFMA.
Delivery: Supplied with ten year warranty.
Spécifications
- Catalog No:
- 76276-878
- 76276-876
- 76276-886
- 76276-884
- 76276-882
- 76276-880
- 76276-892
- 76276-890
- 76276-888
- Model:
- 76276-878
- 76276-876
- 76276-886
- 76276-884
- 76276-882
- 76276-880
- 76276-892
- 76276-890
- 76276-888