A propos de cet article
Connect any ProDIGITAL integrated probe/cable assembly the ProSolo to form a complete sampling system. Probe assemblies are user-replaceable with cable lengths ranging from 1 to 100 m.
- Dissolved oxygen technology has advanced from electrochemical sensors, such as polarographic and galvanic to luminescent-based optical sensors
- Digital smart sensors
- Single cable design for probe assembly lengths ranging from 1 to 100 m
The ODO®/T probe assemblies pair with the ProSolo Handheld to form a complete sampling system. It features integral, digital sensors, the ODO®/T is plug-and-play which minimizes time spent on instrument setup. Pro DIGITAL technology allows the handheld to automatically recognize connected sensors, so there is no need to tell the instrument what is connected.
Optical Dissolved Oxygen (ODO®) technology is non-consumptive, meaning oxygen is not consumed during the measurement. Flow dependence is non-existent and no stirring is required, resulting in the most accurate DO measurements available from a probe. The ODO® sensor is not susceptible to interferences from other gases, drifts less and holds calibration longer than membrane covered sensors. Maintenance is a breeze with a simple, threaded optical cap that is good for at least two years.
Instrument setup is quick and easy with the integral sensors, which eliminates the need to install sensors before use. The digital technology allows the handheld to automatically recognize connected sensors, so there is no need to tell the instrument what is connected.
Spécifications
- Catalog No:
- 76308-040
- 76308-036
- 76308-038
- 76308-358
- 76308-034
- 76308-360
- 76308-042
- DO concentration:
- 0 - 50 mg/l
- 0 - 50 mg/l
- 0 - 50 mg/l
- 0 - 50 mg/l
- 0 - 50 mg/l
- 0 - 50 mg/l
- 0 - 50 mg/l
- DO saturation:
- 0 - 500%
- 0 - 500%
- 0 - 500%
- 0 - 500%
- 0 - 500%
- 0 - 500%
- 0 - 500%
- DO accuracy:
- 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
- 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
- 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
- 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
- 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
- 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
- 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
- DO resolution:
- 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)
- 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)
- 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)
- 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)
- 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)
- 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)
- 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)