LED Head Light, Portwest
# de catalogue 76104-506
Fournisseur: Portwest
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Color:Silver/Black
- Size:Adjustable
Spécifications
A propos de cet article
Our high-quality LED Head Light has tilt control for different working positions.
- 40 Lumen brightness
- 15 yards (high), 10 yards (medium) beam distance
- Ten hours run time
- High-medium-flash function
- 8 LED
Delivery: Supplied with three AAA batteries.
Customers Who Bought This Also Bought