A propos de cet article
CapTrack™ is the ideal benchtop solution for increased efficiency, sample integrity and safety in blood processing labs.
- CT1: Automated tube recapping, tracking and sample management with tube height and cap color recognition recall function
- CT2: Automated tube recapping, tracking and sample management with rack position recall function
- CR1: Automated tube recapping
CapTrack™ automation eliminates the need for time-consuming manual processing that can result in reduced efficiency and increased risk of sample contamination. It gives technicians the freedom to put their professional skills to better use elsewhere in the lab. With CapTrack™ in the lab, the risk of exposure to blood borne pathogens is drastically reduced, as is the potential for repetitive strain injury.