VWR® Autoclavable Letterhead
Supplier: VWR International
Certificates
About this item
Designed for use in autoclaves, this paper can be sterilized and not lose any resolution of printed images.
- Customer name printed in center
- Optional 2-holes drilled at top of sheet
VWR® autoclavable letterhead is ideally suited for applications requiring water resistance and the ability to be written on when wet. This includes use in an autoclave.
Customers Who Bought This Also Bought