OIML Class 4 Individual Precision Weights, ASTM, Mettler Toledo
Fournisseur: Mettler Toledo
Certificats
A propos de cet article
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
- 316 stainless steel
- Low magnetic permeability
- High corrosion resistance
- High gloss polish
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Customers Who Bought This Also Bought