ContactUsLinkComponent
Fluoride Test Papers, CTL Scientific Supply
Fluoride Test Papers, CTL Scientific Supply
# de catalogue 75996-722
Fournisseur:  Macherey-Nagel
Fluoride Test Papers, CTL Scientific Supply
# de catalogue 75996-722
Fournisseur:  Macherey-Nagel
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    Box of 200 Strips
Spécifications
A propos de cet article
Used for the determination of fluoride ions and gaseous hydrogen fluoride (H2F2).
  • Pinkish-red paper turns yellowish-white in the presence of fluorides
  • Sensitivity limit 20 mg/L fluoride
Substantial amounts of chlorates, bromates and sulphates cause white discolorations of the paper. These interferences are readily circumvented.
Customers Who Bought This Also Bought