ContactUsLinkComponent
Exel International Brand Quality Curved Tip Syringes, Air-Tite Products
Exel International Brand Quality Curved Tip Syringes, Air-Tite Products
# de catalogue 76290-492
Fournisseur:  Air-Tite
Syringe Curved Tip Exel Non-St 12Ml BX50
Exel International Brand Quality Curved Tip Syringes, Air-Tite Products
# de catalogue 76290-492
Fournisseur:  Air-Tite
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    12 ml
  • Description:
    Exel Curved Tip Syringe
  • Cat. no.:
    76290-492
  • Packaging:
    50/box
  • Type:
    Curved dispensing tip
Spécifications
A propos de cet article
EXEL international's quality disposable curved tip dispensing syringe. Great value at reasonable costs.
  • Standard three part syringe
  • Latex and pyrogen free
  • Non-toxic
  • Positive plunger stop
EXEL international is known for unsurpassed quality at a low price. The design includes bold print, clear barrels, and siliconized gaskets for smooth, consistent plunger motion and a positive plunger stop.
Packaging: Non sterile.
Customers Who Bought This Also Bought