Chemware® PFA Test Tubes, Saint-Gobain Performance Plastics
These unbreakable test tubes are excellent for trace metal analysis.
- Made from fluoropolymer PFA resin
- Low metal ion content
- Inert
They are non-contaminating, and transparent for easy viewing of contents. They withstand extreme temperature. Can be sterilized repeatedly.
Frequently Bought Together