Interchangeable Glass Syringe with Glass Tip, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Interchangeable syringe featuring a glass tip, permanently fused graduations, and a colored plunger end for easy and accurate dosage measurement.
- Made of high-quality borosilicate glass
- Reinforced flanges that are flat on two sides to prevent rolling
- Marked for non-medical use; no medical license required
Manufactured to ensure the perfect mating of barrels and plungers, so any interchangeable plunger of a particular size will fit any interchangeable barrel of the same size.
Customers Who Bought This Also Bought