Glass Syringes with Luer Lock Tip, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Serial matched syringe featuring a Luer lock tip, permanently fused graduations, and a colored plunger end for easy and accurate dosage measurement.
- Made of high-quality borosilicate glass
- Reinforced flanges that are flat on two sides to prevent rolling
- Marked for non-medical use; no medical license required
Barrels and plungers are serialized and lapped together for a fit which will withstand higher temperatures.
Customers Who Bought This Also Bought