Cryogenic Transfer Hose, Airgas
# de catalogue 47752-588
Fournisseur: Airgas
Spécifications
- Length:4 ft.
- Description:Cryogenic transfer hose, CGA-295׳/₈"MNPT
- Cat. no.:47752-588
- Supplier No.:Y154CH429538
Spécifications
A propos de cet article
Hose features stainless steel anti-kink armor casing.
- Suitable for helium, nitrogen and argon
CGA-end features a 90° bend for ease in connecting to liquid cylinders.
Customers Who Bought This Also Bought