DURAN® Gas Washing Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Autoclavable
- Minimal thermal expansion giving relatively high resistance to temperature changes
Wash bottle with DIN GL 45 screw pitch and adjustable trim with screw cap and two PP knobs with bolting.
Customers Who Bought This Also Bought